Imprimer
Cap sealing mats for VersiPlate 96-well PCR strip tube plates, VersiCap™ Mats
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
AB-1815/100 AB-1820 AB-1820/100 AB-1810/100 AB-1810 AB-1815
732-2595EA 205 EUR
732-2595 732-2596 732-2597 732-2593 732-2592 732-2594
Cap sealing mats for VersiPlate 96-well PCR strip tube plates, VersiCap™ Mats
Scellés Microplaque Sceaux
VersiCap™ mats are designed to seal VersiPlate 96-well PCR strip tubes, but can be used will all Thermo Scientific strip tubes and 96-well plates.
  • Reduces sample set up time by sealing PCR strips rather than sequentially with individual caps
  • Easier to apply uniformly than adhesive seals
  • Available in three different formats - domed or flat caps for PCR in various thermal cyclers, or an optical version for qPCR