Imprimer
VWR®, Tubes PCR, 0,2 ml
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
732-0547EA 85.9 EUR
732-0547 732-0548
VWR®, Tubes PCR, 0,2 ml
Tubes Tubes PCR
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.