Imprimer
Fully-electonic rotary microtome, 3006 EM
Fournisseur: PFM Medical GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0300610
720-2972EA 31200 EUR
720-2972
Fully-electonic rotary microtome, 3006 EM
Microtomes et groupes de microtomes Microtomes
The high performance fully-electronic rotary microtome 3006 EM is suitable for all applications in routine use, research and industry. Excellent section quality, safe and simple operation and easy cleaning and care describe this microtome. The handwheel of Rotary 3006 EM has a removable handle to ensure maximum user safety during motorised sectioning. Additionally, the device can be stopped immediately by pushing the emergency stop button.
  • Ergonomic design with backlash- and maintenance-free horizontal and vertical cross-roller bearing system
  • Smooth-
En savoir plus