Imprimer

Ammonium chloride-[¹⁵N], Overnight Express™, Sigma-Aldrich®

Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Ammonium-15N chloride, Ammonium chloride-15N

71759-4
71759-4EA 2160 EUR
71759-4
Ammonium chloride-[¹⁵N], Overnight Express™, Sigma-Aldrich®
Ammonium chloride-[¹⁵N]

Overnight Express™ Autoinduction NMR Medium, 5 L kit

Formule: H₄ClN
Numéro MDL: MFCD00075417
Numéro CAS: 39466-62-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR