Imprimer
pH indicator papers, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2600-102A 2600-100A 2600-101A 2600-103A 2611-628 2600-104A
700-9753EA 30.3 EUR
700-9753 700-9751 700-9752 700-9754 WHAT2611-628 700-9755
pH indicator papers, Whatman™
Papiers test Bandelettes indicatrices de pH
Can be used for the rapid determination of pH values and has led to many applications in laboratories and industry. Available as standard or three colour ranges as rolls in a handy dispenser.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use
Three colors roll: The paper has three separate indicator dye colour bands. The individual combination of colour change resulting from each test is compared with the colour-coded comparison chart printed on the dispenser, giving improved speed andEn savoir plus