Imprimer
pET-14b DNA, Novagen®
Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
69660-3
69660-3EA 356 EUR
69660-3
pET-14b DNA, Novagen®
Vecteurs, plasmides et librairies
The pET-14b vector carries an N-terminal His·Tag® sequence followed by a thrombin cleavage site and three cloning sites.
  • Packed in a plastic ampoule
  • Store at ≤–70 °C
Informations de livraison: The pET vectors are supplied as purified plasmid DNA (10 µg). It includes an induction control strain (supplied as a glycerol stock).
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable