Imprimer
Orion™ Versa Star™ Pro™ Multi-Parameter Meter, Bench
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VSTAR00
665-0283EA 1250 EUR
665-0283
Orion™ Versa Star™ Pro™ Multi-Parameter Meter, Bench
Compteurs électrochimiques pH-mètres et multi-paramètres
This meter offers interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.
  • Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
  • Auto-read to hold a stable reading; ten password-protected methods per channel
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position
En savoir plus