Imprimer

Éthanol 80% volume, Molecular Biology Grade

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Alcool éthylique

6624.5000EA 111.6 EUR
6624.5000 6624.0500
Éthanol 80% volume, Molecular Biology Grade
Éthanol

Ready to use ethanol 80% in water – pre-qualified for molecular biology applications.


  • 80% ethanol
  • DNase-/RNase- and protease-free
  • Ready to use to avoid dilution in water (save time and avoid contamination)
  • Suitable for the purification and precipitation of DNA, RNA and proteins


Ethanol 80% is required in many protocols (automated or manual) for nucleic acid purification using columns or magnetic bead-based technology. Ethanol is free from DNase/RNase so can be used to purify high quality nucleic acid from a range of sample types.

Formule: C₂H₅OH
Poids moléculaire: 46.07 g/mol
Point d'ébullition: 78.3
Point de fusion: -117
Densité: 0,86 g/cm³ (20 °C)
Point éclair: 12
Température de stockage: Température ambiante
Numéro MDL: MFCD00003568
Numéro CAS: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
Indice Merck: 12,03806

Commander maintenant

Résultats des tests de spécification

Assay (V/V) (ethanol) 79 - 81 %
Density (20/4) 0,856 - 0,862
DNAses (exo-endonucleases) Not detected
RNAses Not detected
Proteases Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR