Imprimer
Orion™ Green Combination pH Electrode
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
GD9156BNWP
662-2536EA 141 EUR
662-2536
Orion™ Green Combination pH Electrode
Électrodes Électrodes pH
This pH combination electrode is one of the first lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.
  • Refillable liquid filling
  • Double-junction for dirty-water testing
  • Duarable epoxy body
  • No disposal issues, no disposal cost
  • Waterproof BNC connector fits any meter with BNC input
Informations de livraison: Supplied with 1 m cable.