Imprimer
Orion™ Pro Star PD216 pH/Ion/DO Bench Meters
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
PSTAR2160 PSTAR2165 PSTAR2167 PSTAR2166
662-2521EA 1332 EUR
662-2521 662-2522 662-2524 662-2523
Orion™ Pro Star PD216 pH/Ion/DO Bench Meters
Compteurs électrochimiques pH-mètres et multi-paramètres
The Thermo Scientific™ Orion™ Pro Star PD216 pH/ion and dissolved oxygen bench meter is designed to provide accurate, reliable electrochemistry testing. Featuring an intuitive interface, enhanced data reporting and robust functionality, the Orion™ Pro Star PD216 meter delivers advanced performance in a modern and simplified package.
  • Large 7" high-resolution colour display
  • Responsive capacitive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps
En savoir plus