Imprimer
Solutions de remplissage, Orion™
Marques : Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
900017 900067 900010 900012 900046 900061 900062 900063 900065 950202 951202 951209
662-0755EA 113 EUR
662-0755 ORIO900067 662-0752 ORIO900012 662-0760 662-0761 662-0762 662-0763 662-0764 662-0850 662-0855 ORIO951209
Solutions de remplissage, Orion™
Électrolytes Solutions de remplissage d'électrode
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
  • Enhance the performance and extend the life of ion selective electrodes
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective combination electrodes