Imprimer
Combination pH electrodes, semi-micro, Orion™ 8103BN ROSS™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
8103BN
662-0722EA 366 EUR
662-0722
Combination pH electrodes, semi-micro, Orion™ 8103BN ROSS™
Électrodes Électrodes pH
These pH electrodes with semi-micro tip are ideal for use in clinical, pharmaceutical, and food labs, wherever sample size is a constraint. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration
En savoir plus