Imprimer
Dissolved oxygen probes, TriOxmatic® 700
Fournisseur: WTW ONLINE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
201670 201672
662-0508EA 2690 EUR
662-0508 662-0286
Dissolved oxygen probes, TriOxmatic® 700
Électrodes Sondes à oxygène dissous
Potentiostatic D.O. sensors with integrated temperature sensor.
  • Integrated lightning protection
  • Sensor monitoring function (SensCheck)
  • Zero current free electrode system
  • Exchangeable membrane head with robust 50 um special membrane
  • Lowest incident flow required
  • Double thermistor system for temperature compensation
  • OxiCal air calibration without accessories
Design according to the minimum requirements defined by ATV (Waste water association) in bulletin M 256 for meters to be used at waste water treatment plants.
To be connected to the EcoLine® orEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable