Imprimer
Portoir pour lames
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
H445570000
631-9109EA 214 EUR
631-9109
Portoir pour lames
Porte-lames
Polystyrene holder for up to 12 microscope slides designed for preparation or staining.
  • Inclined ribs for holding slides on their ends
  • Robust and durable white plastic design
  • Safe storage with visible hinged cases
Cdt: Sold by six items.