Imprimer
Upright microscope, DM500
Fournisseur: LEICA MICROSYSTEMS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
13613215_P4 13613215_P1 13613215_P2 13613215_P3
630-3276EA 3440 EUR
630-3276 630-3273 630-3274 630-3275
Upright microscope, DM500
Microscopes
Leica DM500 binocular microscopes were specifically developed to facilitate science teaching. The DM500 is ideal for entry level life science courses.
  • Leica EZLite™ LED illumination provides a cool white light at 80% lower energy consumption
  • LED illumination also eliminates the need to change any bulbs
  • Pre-centred, pre-focused Abbe condenser eliminates the need for adjustments
  • Student friendly slide holder EZGuide™ helps prevent slide chipping
  • Stage with rounded edges and wear-resistant surface
  • Integrated vertical handle provides easy carrying and lifting
En savoir plus