Imprimer
Systèmes d'illumination LED, EasyLED
Fournisseur: SCHOTT AG Lighting and imaging
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
600101 600121 600121-SETC 600121-SETSA
630-2662EA 511 EUR
630-2662 630-2663 630-3491 630-3492
Systèmes d'illumination LED, EasyLED
Éclairages Éclairage LED
The SCHOTT EasyLED portfolio for stereo microscopy delivers precise LED light for a wide variety of applications, such as laboratory and diagnostics, industrial applications, education, or forensic science. The series offers robust design and easy handling in a compact device, with a range of accessories for specific requirements.
  • Powerful luminance; Intense light output for specialised tasks
  • Maintenance-free LED lifetime of 50000 hours
  • Color temperature: approximately 5600 K
  • Low power consumption; smart engineering leads to optimized energy use
  • Compatible
En savoir plus