Imprimer
Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
Marques : Merck Millipore Calbiochem
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
625718-1
625718-1EA 510 EUR
625718-1
Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
Tampons Tampons biologiques
This buffer solution is used for the purification of chemically synthesised oligonucleotides by HPLC.