Imprimer
Bandelettes indicatrices de température
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0646.1065 0646.1071 0646.1077 0646.1082 0646.1110 0646.1121
620-9151EA 34.3 EUR
620-9151 620-9152 620-9153 620-9154 620-9159 620-9161
Bandelettes indicatrices de température
Bandelettes indicatrices de température
testoterm measuring points are self-adhesive, temperature-resistant films with heat-sensitive elements for monitoring a defined maximum temperature.
  • Irreversible change in colour within two seconds
  • Practical pack with 50 thermometer strips
  • One temperature level
L×P : 15×14 mm