Imprimer
pH food electrode
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0650.0245
620-3662EA 308 EUR
620-3662
pH food electrode
Électrodes Électrodes pH
Robust and dirt-insensitive pH penetration electrode without temperature sensor.
  • With hole diaphragm
  • Unbreakable plastic coating around glass electrode
  • Maintenance-free food-safe gel electrolyte
Ideal for use with Testo 230 instrument.
Informations de livraison: pH electrode, including fixed cable with BNC plug and wetting cap.