Imprimer
WiFi temperature and humidity data loggers, new Saveris 2
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0572.2031 0572.2032 0572.2033 0572.2034 0572.2035
620-2695EA 200 EUR
620-2695 620-2696 620-2697 620-2698 620-2699
WiFi temperature and humidity data loggers, new Saveris 2
Enregistreurs de données
The Saveris 2 WiFi data logger system is the modern solution for monitoring temperature and humidity value in storerooms and work rooms. Installing the system is child's play, and can be carried out either via a browser, or with the Saveris 2 app.
  • Data transfer by wireless LAN
  • All measurement data always available, anywhere, on any instrument
  • Alarms when limit values are violated
  • With Saveris 2 app for easier configuration, push alarms and WiFi range analysis
  • Cost-free online data store (cloud)
  • The long battery life additionally ensures that the Saveris 2
En savoir plus