Imprimer
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
61768-1ML
61768-1MLEA 224 EUR
61768-1ML
Lectin from Triticum vulgaris, immobilized on Agarose CL-4B, for affinity chromatography, Supelco®
Protéines purification Protein Purification Resins
50% aqueous suspension in 1 M NaCl.
  • Capacity: 0,1 to 1,0 mg/ml, packed gel capacity (ovomucoid)
Application: Protein isolation from bordetella pertussis that facilitates entry of calmodulin-sensitive adenylate cyclase into animal cells.
Attention: Storage temperature: Refrigerator.