Imprimer
Coupelles à immersion
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VF2071 VF2072 VF2073 VF2074 VF2075 VF2077
616-3412EA 199 EUR
616-3412 616-3413 769-1144 616-3414 616-3415 616-3417
Coupelles à immersion
Viscosimètres Coupes à écoulement
Dip cups can be used to provide a quick 'break-thread' flow time comparative measurements on site in a manufacturing environment.
  • Titanium anodised aluminium with fixed stainless steel nozzle and handle
  • Engraved with a unique serial number
  • Easy to clean
Certifications: DIN 53211
Informations de commande: The calibration certificate needs to be ordered separately.
Informations de livraison: Stand is not included.