Imprimer
ep Dualfilter T.I.P.S.® Filter Pipette Tips, BioBased Reloads
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0030081.030 0030081.048 0030081.056 0030081.064 0030081.072 0030081.080 0030081.099 0030081.102
613-7262EA 195 EUR
613-7262 613-7263 613-7264 613-7265 613-7266 613-7267 613-7268 613-7269
ep Dualfilter T.I.P.S.® Filter Pipette Tips, BioBased Reloads
Pointes de pipettes
Eppendorf ep Dualfilter T.I.P.S.® BioBased Reloads offer eco-conscious pipetting with tips made from 90% plant-based material, 'PCR clean' and 'Sterile' grades with dual-layer filtration, ensuring unparalleled precision and contamination prevention in sustainable, reloadable formats.
  • Improve the laboratory carbon footprint with ep Dualfilter T.I.P.S.® BioBased in the new Eppendorf Reloads
  • Produced from recycled feedstock in second generation, derived solely from waste and residue streams from vegetable oil production
  • Significantly reduce laboratory waste
En savoir plus