Imprimer
VWR®, Wide Bore Pipette Tips
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
613-7089EA 26.5 EUR
613-7089 613-7090 732-3909 613-7091 732-3910 613-7092 613-7093 732-3911
VWR®, Wide Bore Pipette Tips
Pointes de pipettes
Larger orifice prevents cell fragmentation due to shearing.
  • Ideal for pipetting of macromolecules and viscous samples
  • Ideal for NGS protocol
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.