Imprimer
VWR® LTS Compatible Adjustable Volume Pipettors
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
613-6918EA 1200 EUR
613-6918 613-6919 613-6920 613-6921 613-6922 613-6923 613-6924 613-6925 613-6926 613-6927 613-6928 613-6929 613-6930 613-6931 613-6932
VWR® LTS Compatible Adjustable Volume Pipettors
Pipettes Pipetteurs
The VWR® LTS compatible pipettors reduce ejection force with a unique positive stop technology. This technology along with lighter plunger springs, a durable light weight design, and a comfortable handle design with perfect symmetry makes this the best choice for a lab trying to improve ergonomics.
  • Fully compatible with Rainin LTS tips
  • Lighter springs give a more comfortable and controlled draw and dispense
  • Constructed with durable lightweight materials
  • The ejector is cushioned to reduce dynamic stress on your thumb
  • Precision piston design produces
En savoir plus