Imprimer
ep Dualfilter T.I.P.S.® 384 Racks - Filter Pipette Tips
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0030078.853 0030078.861
613-6807EA 900 EUR
613-6807 613-6808
ep Dualfilter T.I.P.S.® 384 Racks - Filter Pipette Tips
Pointes de pipettes
Eppendorf ep Dualfilter T.I.P.S.® 384 Racks deliver cutting-edge filter pipette tips in 'PCR clean and Sterile' grade, specifically designed for 384-well plate applications, ensuring utmost precision and contamination protection in high-throughput labs. ep Dualfilter T.I.P.S.® 384 pipette tips build a system together with the Research® plus and Xplorer® plus 16- and 24-channel pipettes as well as 20 and 100 µl Move It® adjustable tip spacing pipettes for utmost safety and reliability.
  • Innovative SOFTattach technology allows for reliable tip fit and
En savoir plus