Imprimer
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0030078.691 0030078.705 0030078.713 0030078.721 0030078.730 0030078.748
613-6791EA 207 EUR
613-6791 613-6792 613-6793 613-6794 613-6795 613-6796
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Pointes de pipettes
Eppendorf ep Dualfilter T.I.P.S.® SealMax® Racks provide advanced self-sealing pipette tips in 'PCR clean and Sterile' grade, featuring unique two-phase filters for unparalleled protection against contamination in sensitive lab applications.
  • Reliable protection from liquids, aerosols, and biomolecules
  • No PCR inhibiting additives
  • Protection of pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity
En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Certification environnementale Renewable Energy Certificate, ISO 14001