Imprimer
Combitips® advanced Forensic DNA Grade - Individually Blister-Wrapped Positive Displacement Pipette Tips
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0030089.855 0030089.863 0030089.871
613-5797EA 154 EUR
613-5797 613-5798 613-5799
Combitips® advanced Forensic DNA Grade - Individually Blister-Wrapped Positive Displacement Pipette Tips
Pointes de pipettes
Eppendorf Combitips® advanced in 'Forensic DNA Grade' offer unmatched precision and safety, individually blister-wrapped for maximal DNA contamination prevention. Ideal for forensic applications requiring the utmost accuracy and sterility.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Combitips® advanced in ‘Forensic DNA Grade’ quality conform to the requirements of ISO 18385
En savoir plus