Imprimer
Rechargeable NiMH Battery
Fournisseur: DRUMMOND
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
4-000-035
613-4252EA 90.8 EUR
613-4252
Rechargeable NiMH Battery
Piles
Replacement battery for Drummond Scientific Pipet-Aid® controllers.
NiMH 8.4v 200mAh for:
Portable Pipet-Aid® and Pipet-Aid® XP
3/VH 700AAA 3.6v 750mHh for:
Pipet-Aid® XP2 and Pipet-Aid® XL