Imprimer
VWR®, Serological Pipettes, Triple-Bagged
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
612-5890EA 74.25 EUR
612-5890 612-5891 612-5892 612-5893 612-5894 612-5895 612-5896
VWR®, Serological Pipettes, Triple-Bagged
Pipettes Pipettes sérologiques
Pipettes are individually packed in plastic/plastic and then put in double-bags.
  • Polyolefin filter
  • Latex-, DNase- and RNase-free, non pyrogenic, non cytotoxic
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Dispenser box: Easy opening
Colour coding for easy identification.
Certifications: Manufactured in a Class 100 000 cleanroom environment. Meets BSE/TSE requirement. GPPS compliant USP Class VI.