Imprimer
VWR®, Aspirating Pipettes
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
612-5883EA 55.4 EUR
612-5883 612-5884 612-5885 612-5886 612-5887
VWR®, Aspirating Pipettes
Pipettes Pipettes d'aspiration
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Use at –20 to +50 °C
  • Sterilised by gamma irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf life of 4 years after month of production
  • Free of detectable DNase and RNase
  • Non pyrogenic, non cytotoxic
For aseptically aspirating liquid by using vacuum suction.
Certifications: Products are manufactured under the ISO 13485 standard. Meet the requirements of BSE/TSE.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable