Imprimer
Pipette bulbs
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
19195
612-1939EA 271 EUR
612-1939
Pipette bulbs
Pipettes Poires pour pipettes
Pipetting bulb made of silicone (translucent white).
  • Designed for pipetting with plastic and glass pipettes
  • Silicone has a thermal and chemical resistance superior to latex