Imprimer
Pipettes jaugées, BLAUBRAND®, un trait, classe AS
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
29701 29702 29703 29704 29705 29706 29707 29708 29709 29710 29711 29712 29713 29714 29715 29716 29717 29718 29719
612-1201EA 14.6 EUR
612-1201 612-1202 612-1203 612-1204 612-1205 612-1206 612-1207 612-1208 612-1209 612-1210 612-1211 612-1212 612-1213 612-1214 612-1215 612-1216 612-1217 612-1218 612-1219
Pipettes jaugées, BLAUBRAND®, un trait, classe AS
Pipettes Pipettes jaugées
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time: 5 seconds
  • Batch certificate
  • Also available (on request) with an individual certificate or DAkkS calibration certificate
  • USP version available
DIN EN ISO 648
Certifications: DIN EN ISO 648; DE-M marking.