Imprimer
Moisture Analyser, MB92
Fournisseur: OHAUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
30971924
611-5850EA 3680 EUR
611-5850
Moisture Analyser, MB92
Analyseurs Analyseurs d'humidité
High performance moisture analysis with advanced features for the most demanding application.
  • Robust and durable with the new carbon heating technology and an upgraded full colour capacitive touchscreen
  • MB92 is the perfect choice for users demanding a high level of performance and precision
  • Multiple drying profiles, shut off criteria and methods offer great flexibility in method creation
  • Enhanced password protection ensures greater data security and integrity
  • A full glass panel and tool-free heating chamber assembly makes cleaning and maintenance easy
En savoir plus