Imprimer
VWR® Giant-Digits™, Digital Countdown Timer
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
609-0231EA 30.9 EUR
609-0231
VWR® Giant-Digits™, Digital Countdown Timer
Minuteurs Minuteurs numériques
View giant size digits on this bench timer from across the lab. Visible from shelves, in hoods, or perched on instruments. Eliminates 'squinting' to observe alarm time remaining.
  • Giant-Digits™ with 33,8 mm digit height; viewable from 50 feet
  • Countdown and alarms on single channel for ease-of-use
  • Three-button intuitive operation eliminates reading instructions
  • Automatic Bounceback™ Memory for repetitive timing
  • Counts up during alarming indicating elapsed time since alarming
  • Times up to 99 minutes, 59 seconds
  • Settable to one second with one-second resolution
En savoir plus