Imprimer
Supelco® Discovery® Cyano HPLC Columns
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
59357-U40
59357-U40EA 881 EUR
59357-U40
Supelco® Discovery® Cyano HPLC Columns
Colonnes pour chromatographie
Discovery® Cyano columns have low hydrophobicity for rapid elution of hydrophobic analytes with exceptional peak shape for basic and acidic analytes.
  • Suitable for LC/MS application
  • High retention and resolution of polar compounds
  • Compatible with 100% aqueous phases
Stable pH: 2,0 to 8,0
Informations de livraison: Validation packs includes three columns, each from a different lot of bonded phase. Supelguard™ guard cartridge kit includes one cartridge, a stand-alone holder, a piece of tubing, two nuts and ferrules.