Imprimer
Supelco® Ascentis® Express Phenyl-Hexyl HPLC Columns
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
584621-U 584622-U 584609-U 584610-U 584611-U 584612-U 584613-U 584614-U 584615-U 584616-U 584617-U 584618-U 584619-U 584620-U 584623-U
584621-UEA 570 EUR
584621-U 584622-U 584609-U 584610-U 584611-U 584612-U 584613-U 584614-U 584615-U 584616-U 584617-U 584618-U 584619-U 584620-U 584623-U
Supelco® Ascentis® Express Phenyl-Hexyl HPLC Columns
Colonnes pour chromatographie
Ascentis® Express 160 Å Phenyl-Hexyl high-speed, high-performance liquid chromatography columns are based on a 160 Å Fused-Core® particle design.
The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0,5 μm thick porous shell and the small overall particle size of 2,7 μm.
The endcapped, phenyl-hexyl bonded phase provides a stable, reversed phase packing with a pore structure and pore size that isEn savoir plus