Imprimer
Solutions étalons plasmatique de zinc, BAKER INSTRA-ANALYZED®, J.T.Baker®
Marques : Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5791.0100EA 0 EUR
5791.0100
Solutions étalons plasmatique de zinc, BAKER INSTRA-ANALYZED®, J.T.Baker®
Étalons Étalons cationiques Standards de zinc
Single element plasma standards are used for accuracy and easy of calibration, along with precise detection of trace impurities.
Certifications: Traceable to NIST.
Attention: 10 000 µg/ml= (1,00 % w/v)= 10000 mg/L