Imprimer
Supelco® Ascentis® ES Cyano HPLC Columns
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
577306-U 577307-U 577301-U
577306-UEA 807 EUR
577306-U 577307-U 577301-U
Supelco® Ascentis® ES Cyano HPLC Columns
Colonnes pour chromatographie
Ascentis®ES Cyano HPLC columns are optimised to the three terms of the resolution equation: efficiency, retention, and selectivity.
  • High purity, type B silica for inertness, reproducibility, and stability
  • Modern bonding process optimises bonded phase coverage and maximizes stability, while minimising bleed and unwanted secondary interactions
  • Compatible with LC-MS and all of today's sensitive instruments and methods
  • Scalable sensitivity from analytical to preparative
  • High surface area silica for high preparative loading capacity
  • Impurity: <5 ppm metals
En savoir plus