Imprimer
Solid phase extraction disk, ENVI™-18
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
57171
57171.EA 304 EUR
57171.
Solid phase extraction disk, ENVI™-18
Filtres Disques filtrants
Solid phase extraction disks provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples.
  • Reversed phase retention mechanism
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles
Applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides.