Imprimer
Supelco® TraceCERT® Organic Reference Standard, Benzo[ghi]perylene
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
55488-10MG
55488-10MGEA 235 EUR
55488-10MG
Supelco® TraceCERT® Organic Reference Standard, Benzo[ghi]perylene
Étalons Étalons organiques
  • GHS signal: GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P273 - P391 - P501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9851ece455ab01cbfc148f5faa04e607b93f1f9b&vid=9851ece455ab01cbfc148f5faa04e607b93f1f9b&prid=web&clid=SAPDAM