Imprimer
Vydac 218TP C18, HPLC columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
554-7577EA 1160 EUR
554-7577 554-7578 554-7576 218TP510 218TP3410 218TP3215 218TP3415 218TP34 218GD51 218GCC510 218FSK510 218TP5105 218TP5305 218TP5405 218TP5110 218TP5210 218TP5410 218TP51010 218TP5115 218TP5215 218TP53 218GCC1010 218TP104 218TP1010 218TP1022 218GCC1210 218FSK1210 HICH5119751 218TP101510 218TP101522 218TP15202215 218TP152010 218TP152022 218GD104/N 218GD32/N 218GD34/N 218GD52/N 218GD54/N 218GK104/N 218GK32/N 218GK34/N 218GK52/N 218GK54/N 218TP3405
Vydac 218TP C18, HPLC columns
Colonnes pour chromatographie
Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
First generation polymeric C18 phase with unique selectivity. Recommended for smallEn savoir plus