Imprimer
Viscometers, ROTAVISC SBS
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
10007714 10013658 10013672 10013683 10013820 10013822 10013824
531-1464EA 4440 EUR
531-1464 531-1481 531-1484 531-1478 IKAA10013820 IKAA10013822 IKAA10013824
Viscometers, ROTAVISC SBS
Viscosimètres Viscosimètres rotatifs
SBS (Steel Ball Bearing, Suspended) rotational viscometers extend a large part of the application spectrum of the IKA ROTAVISC device family.
  • Easy to use
  • Highest measurement accuracy
  • Infinitely variable speed
Instead of jewel bearing, these devices are equipped with a special ball bearing technology.
They are characterised by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
Whether simple or demandingEn savoir plus