Imprimer
VWR®, Tubes à centrifuger, fond conique
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
525-0606EA 144 EUR
525-0606 525-0611 525-0607 525-0608 525-0609 525-0610 525-0605 525-0604 525-0612
VWR®, Tubes à centrifuger, fond conique
Tubes Tubes à centrifuger
PP, conical, with white HDPE flat cap or plug cap.
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Caps available in two styles: Flat cap with a smooth surface for additional labelling options, or plug style cap for a secure seal when using shakers and horizontal incubators
  • Engraved graduation at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Can be used for storing and freezing samples at temperatures down to –80 °C
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use
En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Certification environnementale ISO 14001/Certificate No.: 04923E00243R0M