Imprimer
Centrifuges, General Purpose Pro CTS Series
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
75009054
521-3510EA 20910 EUR
521-3510
Centrifuges, General Purpose Pro CTS Series
Centrifugeuses Centrifugeuses sur pieds
General Purpose Pro Centrifuges Cell Therapy Systems (CTS™) Series consist of equipment, documentation, and certifications, as well as compliance services that support Good Manufacturing Practice (GMP) requirements. The CTS Series helps you get up and running faster, stay compliant and current with all necessary regulatory certifications, navigate regulatory audits, and stay on schedule, taking cell therapy from discovery to clinical research and commercial manufacturing. The refrigerated 4 L (4×1000 ml) floor standing and bench models are designed to meet the needs of today’s rapid-fireEn savoir plus