Imprimer
Rotors for Pico™ and Fresco™ centrifuges
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
75003424 75003418 75003436 75003440 75003473 75003489 75003465
521-1105EA 771 EUR
521-1105 521-1104 521-1106 521-1107 521-1108 521-1109 521-2153
Rotors for Pico™ and Fresco™ centrifuges
Centrifugeuses Rotors
Increase nucleic acid sample separation by using the Thermo Scientific™ 10×5 ml rotor with ClickSeal™ biocontainment lid.
Maximized micro-volume capacity, G-force up to 20326×g, ClickSeal biocontainment lid shortens retrieval time with one-click opening, compared to screw-on lids, and provides assurance that samples are securely contained.