Imprimer
Filtres pour seringues, Chromatographie ionique Acrodisc®
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
4483 4485 4583 4585 4683 4685 4783 4785 AP-4585 AP-4785
514-4021EA 307 EUR
514-4021 514-4023 514-4039 514-4040 516-7704 516-7705 516-7712 516-7714 514-4233 514-4234
Filtres pour seringues, Chromatographie ionique Acrodisc®
Filtres Filtres-seringue
Optimised to provide the most consistent results when analysing ionic species. Certified for low level extractables detected by conductivity.
Supor® (hydrophilic polyethersulfone); membrane PP housing.
Acrodisc® PSF syringe filters are Zymark and SOTAX Automation Certified to ensure smooth operation and worry-free performance 24 hours a day in automated workstations.