Imprimer
Filtres sans seringue, Mini-UniPrep™, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
UN203NPUORG UN203NPEORG UN503NPUORG UN203NPUGMF UN203NPEAQU UN203NPENYL UN503NPEAQU UN503NPENYL UN503NPEORG UN203APEAQU UN203APEPES UN203APUNYL UN203APUORG UN203NPUAQU UN203NPUDPP UN203NPUNYL UN503NPUAQU UN503NPUDPP UN503NPUNYL US203NPEAQU US203NPEORG US203NPUAQU US203NPUGMF US203NPUNYL US203NPUORG US203NPUPES US503NPEAQU US503NPENYL US503NPEORG US503NPUAQU US503NPUORG US503NPUGMF US503NPUDPP UN203NPERC UN203NPURC UN503NPERC US203APUNYL
514-8075EA 426 EUR
514-8075 516-6343 516-6356 514-8096 514-8089 514-8090 514-8091 514-8099 514-8100 514-8077 514-8080 514-8085 514-8086 514-8094 514-8095 514-8097 514-8102 514-8103 514-8105 514-8108 514-8110 514-8113 514-8115 514-8116 514-8117 514-8118 514-8120 514-8121 514-8122 514-8125 514-8129 514-8127 514-8126 514-0120 514-0123 514-0124 516-0993
Filtres sans seringue, Mini-UniPrep™, Whatman™
Filtres Dispositifs de filtration sans seringue
The Whatman Mini-UniPrep Syringeless Filters combine four products into one syringeless filter solution for efficient chromatography sample preparation for a broad range of applications. They are built for fast and easy high performance liquid chromatography/ultra high performance liquid chromatography sample preparation and analysis.
  • All-in-one filtration device for quick and cost-effective chromatography sample processing
  • Eliminates the need for separate syringes, syringe membrane filters, vials, and septa
  • Designed for use with an UHPLC or HPLC
En savoir plus