Imprimer
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
AP-4910 AP-4912 AP-4913 AP-4915 AP-4916 AP-4918 AP-4919 AP-4921 AP-4917 AP-4914 AP-4920 AP-4916TC
514-1084EA 213 EUR
514-1084 514-1092 514-1086 514-1093 514-1088 514-1094 514-1090 514-1095 514-1219 514-1180 514-1181 514-1456
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Filtres Filtres-seringue
The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
  • Increased surface area for maximising throughput
  • Heat sealed membrane for reduced breakthrough
  • Accurate analysis. HPLC certified for low levels of UV-absorbing extractables
  • Available in 0,2 µm for UHPLC applications and with GxF (Glass Fiber) pre-filter for difficult to filter samples
  • The Acrodisc® One offers a Universal Membrane for all your analytical filtration requirements