Imprimer
Filtres en fibre de verre
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
60010 61630 61631 61632 61633 61635 61638 61652 61655 61663 61664 64798 66075 66078 66085 66088 66198 66209 66211 66213 66215 66217 66224 66226 66227 66559 60127
516-7725EA 136 EUR
516-7725 513-0001 513-0002 516-7789 516-7790 513-0003 516-7792 513-0004 516-7795 516-7796 516-7797 513-0005 513-0007 513-0008 513-0009 516-7834 513-0011 513-0013 516-7863 513-0015 513-0017 516-7868 513-0022 513-0023 516-9016 516-7939 516-9108
Filtres en fibre de verre
Filtres Filtres de fibres de verre
Superior grade glass fibre filters for a variety of air sampling methods. Used in a variety of sample clean-up, prefiltration and analytical testing applications.
Type A/B: Binder-free borosilicate glass microfibres. For diagnostic applications and sample prefiltration. High loading capacity with 2,5 times thicker glass than type A/C.
Type A/C: Binder-free borosilicate glass microfibres. For testing dissolved and suspended solids in wastewater, for cell harvesting applications and prefiltration.
N° A/D: Microfibre de verre borosilicaté exempt de liant. Excellents préfiltres pourEn savoir plus